Assay Design



Gene Transcripts

Gene Symbol
cbr3
Entrez Gene ID
100145008
Gene Name
carbonyl reductase 3
Gene Aliases
-
Chromosome Location
Chr.2: 9364358 - 9367042 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126568.1 NP_001120040.1 2 - 3 432 58
GenBank mRNA BC158350.1 - 2 - 3 432 58
CU075809.1 - 2 - 3 408 58

View Full Product Details