Assay Design



Gene Transcripts

Gene Symbol
mrpl15
Entrez Gene ID
100145126
Gene Name
mitochondrial ribosomal protein L15
Gene Aliases
-
Chromosome Location
Chr.6: 100113281 - 100120567 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126635.1 NP_001120107.1 4 - 5 669 91
GenBank mRNA BC158958.1 - 4 - 5 669 91

View Full Product Details