Assay Design



Gene Transcripts

Gene Symbol
tbxa2r
Entrez Gene ID
100145351
Gene Name
thromboxane A2 receptor
Gene Aliases
-
Chromosome Location
Chr.1: 74607956 - 74624450 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126823.1 NP_001120295.1 2 - 3 1060 86
GenBank mRNA BC160592.1 - 2 - 3 1060 86

View Full Product Details