Assay Design



Gene Transcripts

Gene Symbol
igf3
Entrez Gene ID
100145502
Gene Name
insulin-like growth factor 3
Gene Aliases
igf-3, igf2
Chromosome Location
Chr.7: 103078148 - 103087708 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126946.1 NP_001120418.1 3 - 4 440 76
GenBank mRNA BC161167.1 - 3 - 4 440 76
CR848377.2 - - 493 76

View Full Product Details