Assay Design



Gene Transcripts

Gene Symbol
mettl22
Entrez Gene ID
100145759
Gene Name
methyltransferase like 22
Gene Aliases
c16orf68
Chromosome Location
Chr.9: 35077888 - 35182101 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127130.1 NP_001120602.1 5 - 6 737 82
GenBank mRNA BC161637.1 - 5 - 6 737 82

View Full Product Details