Assay Design



Gene Transcripts

Gene Symbol
capza1
Entrez Gene ID
550078
Gene Name
capping actin protein of muscle Z-line subunit alpha 1
Gene Aliases
cappa1, capz, caz1
Chromosome Location
Chr.2: 74056833 - 74086491 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017324.3 NP_001017324.1 7 - 8 658 74
GenBank mRNA BC067322.1 - - 587 74
BC158140.1 - 7 - 8 658 74
CR848613.2 - 7 - 8 606 74

View Full Product Details