Assay Design



Gene Transcripts

Gene Symbol
fkbp7
Entrez Gene ID
496687
Gene Name
FK506 binding protein 7
Gene Aliases
-
Chromosome Location
Chr.9: 58991539 - 58997617 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114204.1 NP_001107676.1 2 - 3 459 89
GenBank mRNA BC087818.1 - 2 - 3 395 89
BC157309.1 - 2 - 3 459 89

View Full Product Details