Assay Design



Gene Transcripts

Gene Symbol
stard10
Entrez Gene ID
100145047
Gene Name
StAR related lipid transfer domain containing 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683816: 25526 - 42647 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126587.1 NP_001120059.1 5 - 6 751 76
GenBank mRNA BC158445.1 - 5 - 6 751 76

View Full Product Details