Assay Design



Gene Transcripts

Gene Symbol
edc3
Entrez Gene ID
100145060
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
lsm16, yjdc
Chromosome Location
Chr.3: 84546262 - 84604647 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126590.1 NP_001120062.1 3 - 4 862 92
GenBank mRNA BC158473.1 - 3 - 4 862 92

View Full Product Details