Assay Design



Gene Transcripts

Gene Symbol
idnk
Entrez Gene ID
100145063
Gene Name
idnK, gluconokinase homolog (E. coli)
Gene Aliases
c9orf103
Chromosome Location
Chr.1: 121862440 - 121876070 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126592.1 NP_001120064.1 3 - 4 189 90
XM_018090598.1 XP_017946087.1 3 - 4 209 90
GenBank mRNA BC158476.1 - 3 - 4 189 90

View Full Product Details