Assay Design



Gene Transcripts

Gene Symbol
esm1
Entrez Gene ID
100145234
Gene Name
endothelial cell-specific molecule 1
Gene Aliases
-
Chromosome Location
Chr.1: 178979833 - 178990643 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126720.1 NP_001120192.1 1 - 2 400 81
GenBank mRNA BC159310.1 - 1 - 2 400 81

View Full Product Details