Assay Design



Gene Transcripts

Gene Symbol
dnajc8
Entrez Gene ID
100145281
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Gene Aliases
-
Chromosome Location
Chr.2: 53467765 - 53486728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126759.1 NP_001120231.1 3 - 4 256 96
GenBank mRNA BC160408.1 - 3 - 4 256 96

View Full Product Details