Assay Design



Gene Transcripts

Gene Symbol
cyp1d1
Entrez Gene ID
100145341
Gene Name
cytochrome P450 family 1 subfamily D polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.1: 118488824 - 118502138 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126813.1 NP_001120285.2 2 - 3 897 85
XM_012955974.2 XP_012811428.1 2 - 3 970 85
GenBank mRNA BC160554.1 - 2 - 3 897 85
HQ018043.1 - 2 - 3 840 85

View Full Product Details