Assay Design



Gene Transcripts

Gene Symbol
foxred2
Entrez Gene ID
100145474
Gene Name
FAD-dependent oxidoreductase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.4: 105549574 - 105559959 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126926.1 NP_001120398.1 6 - 7 1449 102
XM_018092927.1 XP_017948416.1 6 - 7 1443 102
XM_018092928.1 XP_017948417.1 6 - 7 1558 102
GenBank mRNA BC161108.1 - 6 - 7 1449 102
CR855483.2 - 2 - 3 271 102

View Full Product Details