Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126926.1 | NP_001120398.1 | 7 - 8 | 1694 | 70 |
XM_018092927.1 | XP_017948416.1 | 7 - 8 | 1688 | 70 | |
XM_018092928.1 | XP_017948417.1 | 7 - 8 | 1803 | 70 | |
GenBank mRNA | BC161108.1 | - | 7 - 8 | 1694 | 70 |
CR855483.2 | - | 3 - 4 | 516 | 70 |