Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127159.1 | NP_001120631.1 | 7 - 8 | 1573 | 74 |
XM_012970105.2 | XP_012825559.1 | 7 - 8 | 1658 | 74 | |
XM_012970106.2 | XP_012825560.1 | 7 - 8 | 1624 | 74 | |
XM_012970107.2 | XP_012825561.1 | 7 - 8 | 2435 | 74 | |
GenBank mRNA | BC161784.1 | - | 7 - 8 | 1573 | 74 |