Assay Design



Gene Transcripts

Gene Symbol
plpp5
Entrez Gene ID
100158576
Gene Name
phospholipid phosphatase 5
Gene Aliases
ppapdc1b
Chromosome Location
Chr.3: 120851188 - 120860213 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128006.2 NP_001121478.1 4 - 5 365 81
GenBank mRNA AL888583.2 - 4 - 5 81
BC166252.1 - 4 - 5 81
BC168452.1 - 3 - 4 252 81
CU024953.1 - 3 - 4 81
CX917894.1 - 4 - 5 81
CX928579.1 - 4 - 5 81

View Full Product Details