Assay Design



Gene Transcripts

Gene Symbol
rhov
Entrez Gene ID
100038217
Gene Name
ras homolog family member V
Gene Aliases
arhv, chp, wrch2
Chromosome Location
Chr.8: 94993937 - 95003125 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128659.1 NP_001122131.1 2 - 3 468 70
GenBank mRNA BC167318.1 - 2 - 3 468 70
CU075657.1 - 2 - 3 479 70

View Full Product Details