Assay Design



Gene Transcripts

Gene Symbol
pou5f3.2
Entrez Gene ID
100170181
Gene Name
POU class 5 homeobox 3, gene 2
Gene Aliases
Oct-25, oct-4, oct25, oct3, oct4, otf3, otf4, pou25, pou5f1.1, xoct-25, xoct25
Chromosome Location
Chr.8: 511490 - 515902 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129934.1 NP_001123406.1 3 - 3 1220 65
GenBank mRNA BC167674.1 - 3 - 3 1220 65
BC167702.1 - 3 - 3 1207 65

View Full Product Details