Assay Design



Gene Transcripts

Gene Symbol
ncald
Entrez Gene ID
100170190
Gene Name
neurocalcin delta
Gene Aliases
-
Chromosome Location
Chr.6: 118906891 - 118967437 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129941.1 NP_001123413.1 3 - 4 770 91
GenBank mRNA BC167579.1 - 3 - 4 770 91

View Full Product Details