Assay Design



Gene Transcripts

Gene Symbol
prdm2
Entrez Gene ID
100170624
Gene Name
PR domain containing 2, with ZNF domain
Gene Aliases
humhoxy1, kmt8, mtb-zf, riz, riz1, riz2
Chromosome Location
Chr.7: 77251512 - 77327606 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130385.1 NP_001123857.1 2 - 3 188 59
XM_012966192.2 XP_012821646.1 3 - 4 611 59
GenBank mRNA BC167736.1 - 2 - 3 188 59

View Full Product Details