Assay Design



Gene Transcripts

Gene Symbol
cd3g
Entrez Gene ID
100216101
Gene Name
CD3g molecule
Gene Aliases
cd3-gamma, cd3g/d, cd3gd, t3g
Chromosome Location
Chr.7: 84890232 - 84895373 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142083.1 NP_001135555.1 1 - 2 129 102
GenBank mRNA BC168039.1 - 1 - 2 129 102

View Full Product Details