Assay Design



Gene Transcripts

Gene Symbol
mkrn3
Entrez Gene ID
100216134
Gene Name
makorin ring finger protein 3
Gene Aliases
makorin-3, rnf63, zfp127, znf127
Chromosome Location
Chr.2: 77286555 - 77297014 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142112.1 NP_001135584.1 3 - 4 398 70
GenBank mRNA BC168097.1 - 3 - 4 398 70

View Full Product Details