Assay Design



Gene Transcripts

Gene Symbol
ribc1
Entrez Gene ID
100216275
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
-
Chromosome Location
Chr.8: 2270344 - 2307411 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142228.2 NP_001135700.1 5 - 6 892 94
XM_012968121.2 XP_012823575.1 6 - 7 988 94
XM_012968122.2 XP_012823576.1 6 - 7 989 94
XM_012968123.1 XP_012823577.1 6 - 7 990 94
XM_012968124.2 XP_012823578.1 6 - 7 911 94
GenBank mRNA BC168813.1 - 5 - 6 894 94
CR761888.2 - 6 - 7 1012 94

View Full Product Details