Assay Design



Gene Transcripts

Gene Symbol
mbd2
Entrez Gene ID
100216281
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
dmtase, ny-co-41
Chromosome Location
Chr.1: 190320792 - 190362502 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142234.1 NP_001135706.1 3 - 4 721 66
GenBank mRNA BC168820.1 - 3 - 4 721 66

View Full Product Details