Assay Design



Gene Transcripts

Gene Symbol
eln2
Entrez Gene ID
100217330
Gene Name
tropoelastin 2
Gene Aliases
-
Chromosome Location
Chr.9: 7003838 - 7026531 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142826.2 NP_001136298.1 27 - 28 1146 90
XM_012970193.2 XP_012825647.1 26 - 27 1139 90
XM_012970194.2 XP_012825648.1 26 - 27 1124 90
XM_012970195.2 XP_012825649.1 27 - 28 1166 90
GenBank mRNA CU075851.1 - 27 - 28 1299 90
EU220008.1 - 27 - 28 1148 90

View Full Product Details