Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142826.2 | NP_001136298.1 | 29 - 30 | 1199 | 105 |
XM_012970193.2 | XP_012825647.1 | 28 - 29 | 1192 | 105 | |
XM_012970194.2 | XP_012825648.1 | 28 - 29 | 1177 | 105 | |
XM_012970195.2 | XP_012825649.1 | 29 - 30 | 1219 | 105 | |
XM_012970196.2 | XP_012825650.1 | 26 - 27 | 1108 | 105 | |
XM_012970197.2 | XP_012825651.1 | 26 - 27 | 1108 | 105 | |
GenBank mRNA | CU075851.1 | - | 29 - 30 | 1352 | 105 |
EU220008.1 | - | 29 - 30 | 1201 | 105 |