Assay Design



Gene Transcripts

Gene Symbol
fgf22
Entrez Gene ID
100233215
Gene Name
fibroblast growth factor 22
Gene Aliases
-
Chromosome Location
Chr.1: 86677012 - 86706810 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143924.1 NP_001137396.1 1 - 2 221 89
XM_012957709.2 XP_012813163.1 3 - 4 677 89
GenBank mRNA FJ472324.1 - 1 - 2 221 89

View Full Product Details