Assay Design



Gene Transcripts

Gene Symbol
arl14
Entrez Gene ID
548508
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
-
Chromosome Location
Chr.5: 103286869 - 103288721 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015791.1 NP_001015791.1 1 - 1 549 93
GenBank mRNA BC089738.1 - 1 - 1 549 93

View Full Product Details