Assay Design



Gene Transcripts

Gene Symbol
mterf1
Entrez Gene ID
549783
Gene Name
mitochondrial transcription termination factor 1
Gene Aliases
mterf
Chromosome Location
Chr.6: 23210984 - 23212626 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017029.2 NP_001017029.1 1 - 1 447 92
GenBank mRNA CR762037.2 - 1 - 1 447 92

View Full Product Details