Assay Design



Gene Transcripts

Gene Symbol
cfh
Entrez Gene ID
394826
Gene Name
complement factor H
Gene Aliases
armd4, arms1, cfhl3, fhl1, hf1, hf2, hus
Chromosome Location
Chr.Un|NW_016684160: 4954 - 24900 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203887.1 NP_989218.1 - 1154 95
GenBank mRNA BC063193.1 - - 1154 95
CT030352.1 - - 1154 95

Gene Symbol
LOC105946053
Entrez Gene ID
105946053
Gene Name
complement factor H-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016684958: 3135 - 4406 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012955263.1 XP_012810717.1 - 1154 95

View Full Product Details