Assay Design



Gene Transcripts

Gene Symbol
sid1
Entrez Gene ID
100271892
Gene Name
sister of indeterminate spikelet 1
Gene Aliases
ZEAMMB73_275310
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146067.1 NP_001139539.1 - 986 70
GenBank mRNA BT063357.1 - - 986 70
EU974186.1 - - 941 70
FJ686697.1 - - 941 70

View Full Product Details