Assay Design



Gene Transcripts

Gene Symbol
exg1
Entrez Gene ID
100191390
Gene Name
exoglucanase1
Gene Aliases
ZEAMMB73_814799
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136824.1 NP_001130296.1 - 1047 62
GenBank mRNA BT033466.1 - - 1047 62
BT033823.1 - - 1073 62
BT054961.1 - - 1047 62
BT066424.1 - - 1047 62

View Full Product Details