Assay Design



Gene Transcripts

Gene Symbol
ARF23
Entrez Gene ID
100191784
Gene Name
auxin response factor 23
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137209.2 NP_001130681.2 - 824 103
GenBank mRNA BT033989.1 - - 750 103

View Full Product Details