Assay Design



Gene Transcripts

Gene Symbol
sqs1
Entrez Gene ID
541614
Gene Name
squalene synthase1
Gene Aliases
ZEAMMB73_448616
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111369.1 NP_001104839.1 - 1367 68
GenBank mRNA AB007502.1 - - 1367 68
BT067860.1 - - 1377 68

View Full Product Details