Assay Design



Gene Transcripts

Gene Symbol
crs1
Entrez Gene ID
541870
Gene Name
chloroplast RNA splicing1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111538.1 NP_001105008.1 - 1600 101
GenBank mRNA AF290414.1 - - 1600 101
AF290415.1 - - 1091 101
AY109620.1 - - 1600 101

View Full Product Details