Assay Design



Gene Transcripts

Gene Symbol
aap1
Entrez Gene ID
100286291
Gene Name
acylaminoacyl-peptidase1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159178.1 NP_001152650.1 - 662 79
GenBank mRNA BT085955.1 - - 629 79
EU976635.1 - - 662 79

View Full Product Details