Assay Design



Gene Transcripts

Gene Symbol
sut2
Entrez Gene ID
100280251
Gene Name
sucrose transporter2
Gene Aliases
ZEAMMB73_722694
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001153179.1 NP_001146651.1 - 1421 76
GenBank mRNA AY581895.1 - - 1164 76
BT055860.1 - - 1421 76

View Full Product Details