Assay Design



Gene Transcripts

Gene Symbol
LOC100281351
Entrez Gene ID
100281351
Gene Name
ligatin
Gene Aliases
ZEAMMB73_308600
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001154269.1 NP_001147741.1 - 545 70
GenBank mRNA BT062039.1 - - 506 70
EU956416.1 - - 545 70

View Full Product Details