Assay Design



Gene Transcripts

Gene Symbol
phot1
Entrez Gene ID
541663
Gene Name
blue-light receptor phototropin 1
Gene Aliases
nph1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111416.1 NP_001104886.1 - 1179 63
GenBank mRNA AF033263.1 - - 1179 63
AY109754.1 - - 1179 63

View Full Product Details