Assay Design



Gene Transcripts

Gene Symbol
cys2
Entrez Gene ID
542438
Gene Name
cysteine synthase2
Gene Aliases
Mcysp
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111999.1 NP_001105469.1 - 690 78
GenBank mRNA X85803.1 - - 690 78

View Full Product Details