Assay Design



Gene Transcripts

Gene Symbol
SnrK1
Entrez Gene ID
542687
Gene Name
SNF1-related protein kinase
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112203.1 NP_001105673.1 - 674 66
GenBank mRNA AY486125.1 - - 674 66
BT040546.1 - - 700 66
BT060618.1 - - 700 66

View Full Product Details