Assay Design



Gene Transcripts

Gene Symbol
spp1
Entrez Gene ID
541868
Gene Name
sucrose-phosphatase1
Gene Aliases
ZmSPP1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111536.1 NP_001105006.1 - 134 93
GenBank mRNA AF283564.2 - - 134 93
EU967840.1 - - 203 93

View Full Product Details