Assay Design



Gene Transcripts

Gene Symbol
kip1
Entrez Gene ID
542080
Gene Name
knotted interacting protein1
Gene Aliases
kip
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111714.1 NP_001105184.1 - 607 68
GenBank mRNA AY082396.1 - - 607 68

View Full Product Details