Assay Design



Gene Transcripts

Gene Symbol
mlo1
Entrez Gene ID
542672
Gene Name
barley mlo defense gene homolog1
Gene Aliases
-Mlo1, Zm-Mlo1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112190.1 NP_001105660.1 - 640 67
GenBank mRNA AY029312.2 - - 640 67
AY110401.1 - - 200 67
BT036101.1 - - 745 67

View Full Product Details