Assay Design



Gene Transcripts

Gene Symbol
mlo7
Entrez Gene ID
542673
Gene Name
barley mlo defense gene homolog7
Gene Aliases
-Mlo7, Zm-Mlo7
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112191.1 NP_001105661.1 - 897 90
GenBank mRNA AY029318.1 - - 897 90

View Full Product Details