Assay Design



Gene Transcripts

Gene Symbol
GRF1
Entrez Gene ID
100101535
Gene Name
growth-regulating factor 1-like
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112551.1 NP_001106021.1 - 334 105
GenBank mRNA EF515840.1 - - 334 105

View Full Product Details