Assay Design



Gene Transcripts

Gene Symbol
rph1
Entrez Gene ID
100192964
Gene Name
RNase PH homolog
Gene Aliases
ZEAMMB73_833729
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001138140.1 NP_001131612.1 - 446 100
GenBank mRNA BT035096.1 - - 446 100
EU960828.1 - - 502 100

View Full Product Details