Assay Design



Gene Transcripts

Gene Symbol
adh1
Entrez Gene ID
542363
Gene Name
alcohol dehydrogenase1
Gene Aliases
Adh1-1F, Adh1-1S
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111939.1 NP_001105409.1 - 518 62
GenBank mRNA L23548.1 - - 473 62
X00580.1 - - 518 62

View Full Product Details