Gene Transcripts
- Gene Symbol
- Nkx2-2os
- Entrez Gene ID
- 100313531
- Gene Name
- NK2 homeobox 2, opposite strand
- Gene Aliases
- Gm14112, Nkx2-2as, Nkx2.2AS, OTTMUSG00000015691
- Chromosome Location
- Chr. 2: 147184083 - 147331681 on Build GRCm38
- UniGene ID
- Mm.405189
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030769.2 |
|
5 |
2899 |
|
GenBank mRNA |
AK045921.1 |
- |
5 |
2900 |
|
View Full Product Details